missing translation for 'onlineSavingsMsg'
Learn More

endo-N-Hydroxy-5-norbornene-2,3-dicarboximide, 97%

Product Code. 11484634
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484634 250 g 250g
11464634 10 g 10g
11474634 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484634 Supplier Thermo Scientific Alfa Aesar Supplier No. A13205.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21715-90-2 | C9H9NO3 | 179.18 g/mol

It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

Solubility
Soluble in ethanol

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21715-90-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00065691
InChI Key ZUSSTQCWRDLYJA-UHFFFAOYNA-N
Synonym honb, n-hydroxy-5-norbornene-2,3-dicarboximide, 2-hydroxy-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3 2h-dione, 4-hydroxy-4-azatricyclo 5.2.1.0 2,6 dec-8-ene-3,5-dione, n-hydroxynorborn-5-ene-2,3-dicarboximide, n-hydroxy-5-norbornene-2,3-dicarboxylic imide, 5-norbornene-2, n-hydroxy, 4,7-methano-1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4-hydroxy-4-azatricyclo 5.2.1.0<2,6> dec-8-ene-3,5-dione
PubChem CID 89529
IUPAC Name 4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES ON1C(=O)C2C3CC(C=C3)C2C1=O

Specifications

Melting Point 166°C to 170°C
Quantity 250 g
Beilstein 13540
Solubility Information Soluble in ethanol
Formula Weight 179.18
Percent Purity 97%
Chemical Name or Material endo-N-Hydroxy-5-norbornene-2,3-dicarboximide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.