Learn More
BUCHI End Pistons
Product Code. 12880713 Shop All Buchi Products
Click to view available options
Capacity (Metric):
40 mL
Unit Size:
Each
Product Code. 12880713
Brand: BUCHI 036478
Only null left
Add to basket
Add to basket
Short path distillation accessories for small scale R and D
  • Up to 4 bulbs for fractional distillation
  • 5 to 40 mL sizes
Specifications
Capacity (Metric) 40 mL
Type End Piston