missing translation for 'onlineSavingsMsg'
Learn More

Encorafenib, MedChemExpress

Product Code. 16472757
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16472757

Brand: Medchem Express HY1560550mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Encorafenib (LGX818) is a highly potent BRAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing BRAFV600E (EC50=4 nM).

Encorafenib (LGX818) is a highly potent BRAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing BRAFV600E (EC50=4 nM).

Specifications

Chemical Name or Material Encorafenib
CAS 1269440-17-6
Color White
Purity Grade Notes Research
Molecular Formula C22H27ClFN7O4S
Quantity 50 mg
Synonym LGX818
Solubility Information DMSO : 50 mg/mL (92.59 mM; Need ultrasonic)
SMILES O=C(OC)N[C@@H](C)CNC1=NC=CC(C2=CN(C(C)C)N=C2C3=CC(Cl)=CC(NS(=O)(C)=O)=C3F)=N1
Molecular Weight (g/mol) 540.01
Formula Weight 540.01
Percent Purity 95.12%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.