missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Encequidar mesylate, MedChemExpress
Description
Encequidar mesylate (HM30181 mesylate
Specifications
Specifications
| Chemical Name or Material | Encequidar mesylate |
| CAS | 849675-87-2 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C39H40N6O10S |
| Quantity | 50 mg |
| Synonym | HM30181 mesylateHM30181A mesylate |
| Solubility Information | DMSO : 25 mg/mL (31.85 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC(C2=CC=CC=C2O1)=O)NC3=CC(OC)=C(OC)C=C3C4=NN(C5=CC=C(CCN6CC7=C(C=C(OC)C(OC)=C7)CC6)C=C5)N=N4.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 784.83 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?