missing translation for 'onlineSavingsMsg'
Learn More

Enbucrilate, Technical Grade, TRC

Product Code. 30373190
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373190 5 g
30372258 25 g
30386069 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373190 Supplier Toronto Research Chemicals Supplier No. TRCE5554705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6606-65-1
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
Synonym 2-Cyano-2-propenoic Acid Butyl Ester, 2-Cyano-acrylic Acid Butyl Ester, BK 200, BK 201, BK 201 (adhesive), BK 300, BK 301, BK 301 (adhesive), Butyl 2-cyanoacrylate, Butyl cyanoacrylate, Butyl α-cyanoacrylate, Enbucrilate, Glubranh 1, Histacryl Blue, Hystoacryl, Indermil, Indermil Tissue Adhesive, InteguSeal Clear, Integuseal, Integuseal Microbial Sealant, Sicomet 6000, Tisuacryl, n-Butyl 2-cyanoacrylate
IUPAC Name butyl 2-cyanoprop-2-enoate
SMILES COC(=O)C1C(c2ccccc2)(O1)c3ccccc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H11NO2/c1-3-4-5-11-8(10)7(2)6-9/h2-5H2,1H3
Formula Weight 153.079
Chemical Name or Material Enbucrilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.