missing translation for 'onlineSavingsMsg'
Learn More

Enbucrilate, MedChemExpress

Product Code. 16495076
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16495076 Supplier Medchem Express Supplier No. HY107346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enbucrilate (Butyl cyanoacrylate) is a cyanoacrylate ester that has been used as surgical tissue adhesive.

Enbucrilate (Butyl cyanoacrylate) is a cyanoacrylate ester that has been used as surgical tissue adhesive.

Specifications

Chemical Name or Material Enbucrilate
CAS 6606-65-1
Color Colorless
Purity Grade Notes Research
Molecular Formula C8H11NO2
Quantity 100 mg
Synonym Butyl cyanoacrylate
Solubility Information DMSO : 100 mg/mL (652.83 mM; Need ultrasonic)
SMILES CCCCOC(C(C#N)=C)=O
Molecular Weight (g/mol) 153.18
Formula Weight 153.18
Percent Purity 95.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.