missing translation for 'onlineSavingsMsg'
Learn More

Enasidenib, MedChemExpress

Product Code. 16447588
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447588 10 mg 10mg
16443607 50 mg 50mg
16427516 100 mg 100mg
16400068 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447588 Supplier Medchem Express Supplier No. HY18690
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Enasidenib is an oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes, with IC50s of 100 and 400 nM against IDH2R140Q and IDH2R172K, respectively.

Enasidenib is an oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes, with IC50s of 100 and 400 nM against IDH2R140Q and IDH2R172K, respectively.

Specifications

Chemical Name or Material Enasidenib
CAS 1446502-11-9
Color White
Purity Grade Notes Research
Molecular Formula C19H17F6N7O
Quantity 10 mg
Synonym AG-221
Solubility Information DMSO : 50 mg/mL (105.62 mM; Need ultrasonic)
SMILES CC(O)(C)CNC1=NC(C2=NC(C(F)(F)F)=CC=C2)=NC(NC3=CC(C(F)(F)F)=NC=C3)=N1
Molecular Weight (g/mol) 473.38
Formula Weight 473.38
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.