missing translation for 'onlineSavingsMsg'
Learn More

Enantiomer of Sofosbuvir, MedChemExpress

Product Code. 16481248
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481248 5 mg 5mg
16442469 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481248 Supplier Medchem Express Supplier No. HYI0726
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Enantiomer of Sofosbuvir is an enantiomer of Sofosbuvir, a prescription medicine for the treatment of patients with chronic hepatitis C. There is no biological activity report on enantiomer of Sofosbuvir until now.

Enantiomer of Sofosbuvir is an enantiomer of Sofosbuvir, a prescription medicine for the treatment of patients with chronic hepatitis C. There is no biological activity report on enantiomer of Sofosbuvir until now.

Specifications

Chemical Name or Material Enantiomer of Sofosbuvir
Color White
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 5 mg
SMILES O=[P@](N[C@H](C)C(OC(C)C)=O)(OC[C@H]1[C@H](O)[C@@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.