missing translation for 'onlineSavingsMsg'
Learn More

Enalaprilat, Thermo Scientific™

The active metabolite of enalapril

Brand:  Thermo Scientific Alfa Aesar J63392.MF

Quantity 50mg

 View more versions of this product

Product Code. 15455639

  • ONLINE EXCLUSIVE PRICE
    £321.00
    £313.99 / 50mg
    Save £7.01
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Enalapril is a ACE (angiotensin-converting enzyme) inhibitor, suggested to be more potent than Captopril (sc-200566). Studies indicate that Enalapril downregulates the expression of IL-β, IL-6, and NF-κB. Further studies show that Enalapril upregulates the expression of VEGF and heme oxygenase-1. Enalaprilat reports demonstrate that the compound has some antioxidant activity, which does not include the PKC-NADPH oxidase pathway.

Solubility
Soluble in water with heating (∼60°C)

Notes
Store in a cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

76420-72-9
348.399
LZFZMUMEGBBDTC-QEJZJMRPSA-N
5462501
(2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid
C18H24N2O5
MFCD00865786
enalaprilat, enalapril acid, enalaprilate, enalaprilatum, enalapril diacid, enalaprilic acid, enalaprilat anhydrous, n-1s-1-carboxy-3-phenylpropyl-l-alanyl-l-proline, unii-q508q118jm
CHEBI:4786
CC(C(=O)N1CCCC1C(=O)O)NC(CCC2=CC=CC=C2)C(=O)O

Specifications

76420-72-9
MFCD00865786
14,3568
Soluble in water with heating (∽60°C)
CC(C(=O)N1CCCC1C(=O)O)NC(CCC2=CC=CC=C2)C(=O)O
348.399
CHEBI:4786
Enalaprilat
C18H24N2O5
50mg
enalaprilat, enalapril acid, enalaprilate, enalaprilatum, enalapril diacid, enalaprilic acid, enalaprilat anhydrous, n-1s-1-carboxy-3-phenylpropyl-l-alanyl-l-proline, unii-q508q118jm
LZFZMUMEGBBDTC-QEJZJMRPSA-N
(2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid
5462501
384.42
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.