missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalaprilat-d5 Sodium Salt, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Enalaprilat-d5 Sodium Salt |
| CAS | 1356922-29-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H18D5N2NaO5 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C18H24N2O5.Na/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13;/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25);/q;+1/p-1/t12-,14-,15-;/m0./s1/i2D,3D,4D,6D,7D; |
| SMILES | [Na+].[2H]c1c([2H])c([2H])c(CC[C@H](N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)[O-])C(=O)O)c([2H])c1[2H] |
| Molecular Weight (g/mol) | 375.41 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction