missing translation for 'onlineSavingsMsg'
Learn More

Enalaprilat-d5 Sodium Salt, TRC

Product Code. 30374129
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30374129

Brand: Toronto Research Chemicals TRCE55537710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Enalaprilat-d5 Sodium Salt
CAS 1356922-29-6
Purity Grade Notes HPLC
Molecular Formula C18H18D5N2NaO5
Quantity 10 mg
InChI Formula InChI=1S/C18H24N2O5.Na/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13;/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25);/q;+1/p-1/t12-,14-,15-;/m0./s1/i2D,3D,4D,6D,7D;
SMILES [Na+].[2H]c1c([2H])c([2H])c(CC[C@H](N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)[O-])C(=O)O)c([2H])c1[2H]
Molecular Weight (g/mol) 375.41
Percent Purity >95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.