missing translation for 'onlineSavingsMsg'
Learn More

Enalaprilat Butyl tert-Butyl Diester, TRC

Product Code. 30386412
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386412 5 mg 5mg
30380285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386412 Supplier Toronto Research Chemicals Supplier No. TRCE5554005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Enalaprilat Butyl tert-Butyl Diester
Analyte or Component Names Enalaprilat Butyl tert-Butyl Diester
Shipping Condition Room Temperature
Molecular Formula C26 H40 N2 O5
Synonym N-[(1S)-3-phenyl-1-[butyloxycarbonyl]propyl]-L-alanyl-L-proline 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C26H40N2O5/c1-6-7-18-32-24(30)21(16-15-20-12-9-8-10-13-20)27-19(2)23(29)28-17-11-14-22(28)25(31)33-26(3,4)5/h8-10,12-13,19,21-22,27H,6-7,11,14-18H2,1-5H3/t19-,21-,22-/m0/s1
Solution Type Neat
SMILES CCCCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-1-[(2S)-2-[[(2S)-1-butoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 460.606
Quantity 5 mg
Formula Weight 460.294
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.