missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalaprilat Butyl tert-Butyl Diester, TRC
Specifications
Specifications
| Chemical Name or Material | Enalaprilat Butyl tert-Butyl Diester |
| Analyte or Component Names | Enalaprilat Butyl tert-Butyl Diester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H40 N2 O5 |
| Synonym | N-[(1S)-3-phenyl-1-[butyloxycarbonyl]propyl]-L-alanyl-L-proline 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C26H40N2O5/c1-6-7-18-32-24(30)21(16-15-20-12-9-8-10-13-20)27-19(2)23(29)28-17-11-14-22(28)25(31)33-26(3,4)5/h8-10,12-13,19,21-22,27H,6-7,11,14-18H2,1-5H3/t19-,21-,22-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl (2S)-1-[(2S)-2-[[(2S)-1-butoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?