missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalaprilat Acyl Glucuronide (~70%), TRC
Specifications
Specifications
| Chemical Name or Material | Enalaprilat Acyl Glucuronide |
| Analyte or Component Names | Enalaprilat Acyl Glucuronide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H32 N2 O11 |
| Synonym | (2S,3S,4S,5R,6S)-6-(((S)-1-((S)-2-(((S)-1-Carboxy-3-phenylpropyl)amino)propanoyl)pyrrolidine-2-carbonyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic Acid |
| InChI Formula | InChI=1S/C24H32N2O11/c1-12(25-14(21(31)32)10-9-13-6-3-2-4-7-13)20(30)26-11-5-8-15(26)23(35)37-24-18(29)16(27)17(28)19(36-24)22(33)34/h2-4,6-7,12,14-19,24-25,27-29H,5,8-11H2,1H3,(H,31,32)(H,33,34)/t12-,14-,15-,16-,17-,18+,19-,24-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2CCC[C@H]2C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-[(2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carbonyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?