missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Enalapril maleate salt

 

Brand:  Thermo Scientific™ J63932.06

Quantity 5g

Product Code. 15406099

  • ONLINE EXCLUSIVE PRICE
    £158.00
    £155.00 / 5g
    Save £3.00
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Enalapril maleate salt is used as a long-acting ACE inhibitor and antihypertensive, used to study diabetic angiopathy in diabetic rats and inhibition of ACE in hog plasma (I50=1.2nM). ACE inhibitors disturb the enin-angiotensin- ald
TRUSTED_SUSTAINABILITY
Specifications

Specifications

76095-16-4
Store at -20°C
492.53
5g
White
Powder
Enalapril maleate salt
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.