missing translation for 'onlineSavingsMsg'
Learn More

Enalapril maleate, MedChemExpress

Product Code. 16442139
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442139 1 g 1g
16420888 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442139 Supplier Medchem Express Supplier No. HYB0331A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enalapril (maleate) (MK-421 (maleate)), the active metabolite of enalapril, is an angiotensin-converting enzyme (ACE) inhibitor.

Enalapril (maleate) (MK-421 (maleate)), the active metabolite of enalapril, is an angiotensin-converting enzyme (ACE) inhibitor.

Specifications

Chemical Name or Material Enalapril maleate
CAS 76095-16-4
Color White
Purity Grade Notes Research
Molecular Formula C24H32N2O9
Quantity 1 g
Synonym MK-421 maleate
Solubility Information DMSO : ≥ 50 mg/mL (101.52 mM) ∣H2O : 33.33 mg/mL (67.67 mM; Need ultrasonic and warming)
SMILES O=C(O)[C@H]1N(C([C@H](C)N[C@H](C(OCC)=O)CCC2=CC=CC=C2)=O)CCC1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 492.52
Formula Weight 492.52
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.