missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalapril Maleate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Enalapril Maleate |
| CAS | 76095-16-4 |
| Analyte or Component Names | Enalapril Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H28 N2 O5 . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Enalapril Maleate, L-Proline, N-[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-, (2Z)-2-butenedioate (1:1), L-Proline, 1-[N-[1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl]-, (S)-, (Z)-2-butenedioate (1:1), Acetensil, Alphrin, Amprace, Analept, Antiprex, BQL, Benalipril, Biocronil, Bitensil, Cardiovet, Controlvas, Converten, Convertin, Ednyt, Elfonal, Enacard, Enaladil, Enalapril maleate, Enaloc, Enapren, Enapril, Enaprin, Enaril, Envas, Glioten, Hipoartel, Hytrol, Innovace, Innovade, Inoprilat, Invoril, Kenopril, Lapril, Lotrial, MK 421, MK 421 maleate, Minipril, Naprilene, Naritec, Nuril, Olivin, Pres, Presil, Pressotec, Renavace, Renitec, Renitek, Reniten, Renivace, Repantril, Sintec, Tenace, Unaril, Unipril, Vasopril, Vasotec, Xanef |
| InChI Formula | InChI=1S/C20H28N2O5.C4H4O4/c1-3-27-20(26)16(12-11-15-8-5-4-6-9-15)21-14(2)18(23)22-13-7-10-17(22)19(24)25;5-3(6)1-2-4(7)8/h4-6,8-9,14,16-17,21H,3,7,10-13H2,1-2H3,(H,24,25);1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,16-,17-;/m0./s1 |
| Solution Type | Neat |
| SMILES | CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)O.OC(=O)\C=C/C(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction