missing translation for 'onlineSavingsMsg'
Learn More

Enalapril maleate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312849
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30312849

Brand: Dr. Ehrenstorfer DREA13119000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Enalapril Maleate
CAS 76095-16-4
Analyte or Component Names Enalapril Maleate
Shipping Condition +20°C
Molecular Formula C20 H28 N2 O5 . C4 H4 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Enalapril Maleate, L-Proline, N-[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-, (2Z)-2-butenedioate (1:1), L-Proline, 1-[N-[1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl]-, (S)-, (Z)-2-butenedioate (1:1), Acetensil, Alphrin, Amprace, Analept, Antiprex, BQL, Benalipril, Biocronil, Bitensil, Cardiovet, Controlvas, Converten, Convertin, Ednyt, Elfonal, Enacard, Enaladil, Enalapril maleate, Enaloc, Enapren, Enapril, Enaprin, Enaril, Envas, Glioten, Hipoartel, Hytrol, Innovace, Innovade, Inoprilat, Invoril, Kenopril, Lapril, Lotrial, MK 421, MK 421 maleate, Minipril, Naprilene, Naritec, Nuril, Olivin, Pres, Presil, Pressotec, Renavace, Renitec, Renitek, Reniten, Renivace, Repantril, Sintec, Tenace, Unaril, Unipril, Vasopril, Vasotec, Xanef
InChI Formula InChI=1S/C20H28N2O5.C4H4O4/c1-3-27-20(26)16(12-11-15-8-5-4-6-9-15)21-14(2)18(23)22-13-7-10-17(22)19(24)25;5-3(6)1-2-4(7)8/h4-6,8-9,14,16-17,21H,3,7,10-13H2,1-2H3,(H,24,25);1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,16-,17-;/m0./s1
SMILES CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)O.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;(2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 492.52
Quantity 1 mL
Formula Weight 492.2108 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.