missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalapril Cyclohexyl Analogue Trifluoroacetic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Enalapril Cyclohexyl Analogue Trifluoroacetic Acid Salt |
| Analyte or Component Names | Enalapril Cyclohexyl Analogue Trifluoroacetic Acid Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H34 N2 O5 . C2 H F3 O2 |
| Synonym | (2S)-1-[(2S)-2-[[(1S)-3-Cyclohexyl-1-(ethoxycarbonyl)propyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid Trifluoroacetic Acid Salt, Enalapril Impurity H, N-[(1S)-1-(Ethoxycarbonyl)-3-cyclohexylpropyl]-L-alanyl-L-proline Trifluoroacetic Acid Salt |
| InChI Formula | InChI=1S/C20H34N2O5.C2HF3O2/c1-3-27-20(26)16(12-11-15-8-5-4-6-9-15)21-14(2)18(23)22-13-7-10-17(22)19(24)25;3-2(4,5)1(6)7/h14-17,21H,3-13H2,1-2H3,(H,24,25);(H,6,7)/t14-,16-,17-;/m0./s1 |
| Solution Type | Neat |
| SMILES | CCOC(=O)[C@H](CCC1CCCCC1)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)O.OC(=O)C(F)(F)F |
| IUPAC Name | (2S)-1-[(2S)-2-[[(2S)-4-cyclohexyl-1-ethoxy-1-oxobutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid;2,2,2-trifluoroacetic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?