missing translation for 'onlineSavingsMsg'
Learn More

Enalapril Acyl Glucuronide, TRC

Product Code. 30376432
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376432 1 mg 1mg
30388484 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376432 Supplier Toronto Research Chemicals Supplier No. TRCE5552251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Enalapril Acyl Glucuronide
Analyte or Component Names Enalapril Acyl Glucuronide
Shipping Condition Room Temperature
Molecular Formula C26 H36 N2 O11
Synonym (2S,3S,4S,5R,6S)-6-(((S)-1-((S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)propanoyl)pyrrolidine-2-carbonyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic Acid
InChI Formula InChI=1S/C26H36N2O11/c1-3-37-24(35)16(12-11-15-8-5-4-6-9-15)27-14(2)22(32)28-13-7-10-17(28)25(36)39-26-20(31)18(29)19(30)21(38-26)23(33)34/h4-6,8-9,14,16-21,26-27,29-31H,3,7,10-13H2,1-2H3,(H,33,34)/t14?,16-,17+,18-,19-,20+,21-,26-/m0/s1
Solution Type Neat
SMILES CCOC(=O)[C@H](CCc1ccccc1)NC(C)C(=O)N2CCC[C@@H]2C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6S)-6-[(2R)-1-[2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carbonyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 552.571
Quantity 1 mg
Formula Weight 552.232
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.