missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENAH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26421-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the enabled/ vasodilator-stimulated phosphoprotein. Members of this gene family are involved in actin-based motility. This protein is involved in regulating the assembly of actin filaments and modulates cell adhesion and motility. Alternate splice variants of this gene have been correlated with tumor invasiveness in certain tissues and these variants may serve as prognostic markers. A pseudogene of this gene is found on chromosome 3.Specifications
| ENAH | |
| Polyclonal | |
| Unconjugated | |
| ENAH | |
| ENA, enabled homolog (Drosophila), ENAH, MENA, NDPP1, Protein enabled homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55740 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N8S7 | |
| ENAH | |
| ENAH Fusion Protein Ag24200 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction