missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EN1 Recombinant Rabbit Monoclonal Antibody (3B4K8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537947
This item is not returnable.
View return policy
Description
Positive Samples: SH-SY5Y, Mouse brain, Rat brain Immunogen sequence: KLKKKKNEKE DKRPRTAFTA EQLQRLKAEF QANRYITEQR RQTLAQELSL NESQIKIWFQ NKRAKIKKAT GIKNGLALHL MAQGLYNHST TTVQDKDESE.
EN1 (Engrailed homolog 1) is a transcription factor expressed in embryonic ventral limb ectoderm and is essential for ventral limb patterning. En1 expression in the developing perichordal tube suggests that it may also be involved in vertebrae assembly. Loss of EN1 function in mice results in dorsal transformations of ventral paw structures and in subtle alterations along the proximal/distal limb axis.
Specifications
| EN1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09065 | |
| EN1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3B4K8 | |
| Unconjugated | |
| EN1 | |
| En1; En-1; engrailed 1; engrailed homeobox 1; engrailed homolog 1; engrailed-1; HME1; homeobox protein en-1; Homeobox protein engrailed-1; hu-En-1; Mo-en.1; mo-En-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13798, 685360 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction