missing translation for 'onlineSavingsMsg'
Learn More

EN 12918 Organophosphorus Mixture 440 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30313125
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313125 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313125 Supplier Dr. Ehrenstorfer Supplier No. DREA50000440AC
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azinphos-ethyl∣Azinphos-methyl∣Bromophos-methyl∣Chlorfenvinphos∣Chlorpyrifos∣Chlorpyrifos-methyl∣Diazinon∣Dichlorvos∣Dimethoate∣Fenitrothion∣Fenthion∣Malathion∣Mevinphos∣Parathion-ethyl∣Parathion-methyl∣Phosalone∣Propetamphos∣Triadimefon∣Triazophos
CAS 2642-71-9, 86-50-0, 2104-96-3, 470-90-6, 2921-88-2, 5598-13-0
Analyte or Component Names Azinphos-ethyl∣Azinphos-methyl∣Bromophos-methyl∣Chlorfenvinphos∣Chlorpyrifos∣Chlorpyrifos-methyl∣Diazinon∣Dichlorvos∣Dimethoate∣Fenitrothion∣Fenthion∣Malathion∣Mevinphos∣Parathion-ethyl∣Parathion-methyl∣Phosalone∣Propetamphos∣Triadimefon∣Triazophos
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Azinphos-ethyl 100 μg/mL∣Azinphos-methyl 100 μg/mL∣Bromophos-methyl 100 μg/mL∣Chlorfenvinphos 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Chlorpyrifos-methyl 100 μg/mL∣Diazinon 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Fenitrothion 100 μg/mL∣Fenthion 100 μg/mL∣Malathion 100 μg/mL∣Mevinphos 100 μg/mL∣Parathion-ethyl 100 μg/mL∣Parathion-methyl 100 μg/mL∣Phosalone 100 μg/mL∣Propetamphos 100 μg/mL∣Triadimefon 100 μg/mL∣Triazophos 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.