missing translation for 'onlineSavingsMsg'
Learn More

EN 12766/CEN EN 61619 PCB Calibration Mixture 10 μg/mL in Isooctane, Dr. Ehrenstorfer

Product Code. 30319398
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30319398 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30319398

Brand: Dr. Ehrenstorfer DREGA09000978IO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 37680-73-2, 31508-00-6, 35065-28-2, 38380-04-0, 35065-27-1, 35065-30-6
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.