missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emx1 Antibody, HRP, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41151H
This item is not returnable.
View return policy
Description
Emx1 Polyclonal antibody specifically detects Emx1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Emx1 | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 2016 | |
| Store at 4°C in the dark. | |
| IgY |
| Western Blot, ELISA | |
| HRP | |
| empty spiracles homeobox 1, empty spiracles-like protein 1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human EMX1. The immunogen is located within amino acids 50 - 100 of EMX1. Amino Acid Squence: CAAAASEEPLRPTALN | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction