missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine Sulfoxide, Mikromol™

Product Code. 17971796
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971796 100 mg 100mg
17912386 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971796 Supplier Mikromol Supplier No. MM1325.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Emtricitabine Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Emtricitabine S-Oxide (Mixture of Diastereomers)
CAS 152128-77-3
Analyte or Component Names Emtricitabine S-Oxide (Mixture of Diastereomers)
Shipping Condition Room Temperature
Molecular Formula C8 H10 F N3 O4 S
Certifications/Compliance ISO 9001
Synonym 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-, 1,3-Oxathiolane, 2(1H)-pyrimidinone deriv., 4-Amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone, 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, S-oxide, [2R-(2alpha,5alpha)]-, 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-, [2R-(2alpha,5alpha)]- (9CI)
InChI Formula InChI=1S/C8H10FN3O4S/c9-4-1-12(8(14)11-7(4)10)5-3-17(15)6(2-13)16-5/h1,5-6,13H,2-3H2,(H2,10,11,14)/t5-,6+,17?/m0/s1
SMILES NC1=NC(=O)N(C=C1F)[C@@H]2CS(=O)[C@H](CO)O2
IUPAC Name 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxo-1,3-oxathiolan-5-yl]pyrimidin-2-one
Molecular Weight (g/mol) 263.25
Quantity 100 mg
Formula Weight 263.0376 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.