missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emtricitabine Diphosphate Triethylammonium Salt (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | Emtricitabine Diphosphate Triethylammonium Salt |
| Analyte or Component Names | Emtricitabine Diphosphate Triethylammonium Salt |
| Shipping Condition | Blue Ice |
| Molecular Formula | C8H12FN3O9P2S • x(C6H15N) |
| Synonym | (2R-cis)-Mono[[5-(4-amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolan-2-yl]methyl] Ester Diphosphoric Acid Triethylammonium Salt, 1,3-Oxathiolane, Diphosphoric Acid Deriv. Triethylammonium Salt |
| InChI Formula | InChI=1S/C8H12FN3O9P2S.C6H15N/c9-4-1-12(8(13)11-7(4)10)5-3-24-6(20-5)2-19-23(17,18)21-22(14,15)16;1-4-7(5-2)6-3/h1,5-6H,2-3H2,(H,17,18)(H2,10,11,13)(H2,14,15,16);4-6H2,1-3H3/t5-,6+;/m0./s1 |
| Solution Type | Neat |
| SMILES | CCN(CC)CC.NC1=NC(N([C@@H]2CS[C@H](COP(OP(O)(O)=O)(O)=O)O2)C=C1F)=O |
| IUPAC Name | ((2R,5S)-5-(4-amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolan-2-yl)methyl trihydrogen diphosphate triethylamine |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?