missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine Diphosphate Triethylammonium Salt (~90%), TRC

Product Code. 30379505
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379505 1 mg 1mg
30383343 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379505 Supplier Toronto Research Chemicals Supplier No. TRCE5249781MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Emtricitabine Diphosphate Triethylammonium Salt
Analyte or Component Names Emtricitabine Diphosphate Triethylammonium Salt
Shipping Condition Blue Ice
Molecular Formula C8H12FN3O9P2S • x(C6H15N)
Synonym (2R-cis)-Mono[[5-(4-amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolan-2-yl]methyl] Ester Diphosphoric Acid Triethylammonium Salt, 1,3-Oxathiolane, Diphosphoric Acid Deriv. Triethylammonium Salt
InChI Formula InChI=1S/C8H12FN3O9P2S.C6H15N/c9-4-1-12(8(13)11-7(4)10)5-3-24-6(20-5)2-19-23(17,18)21-22(14,15)16;1-4-7(5-2)6-3/h1,5-6H,2-3H2,(H,17,18)(H2,10,11,13)(H2,14,15,16);4-6H2,1-3H3/t5-,6+;/m0./s1
Solution Type Neat
SMILES CCN(CC)CC.NC1=NC(N([C@@H]2CS[C@H](COP(OP(O)(O)=O)(O)=O)O2)C=C1F)=O
IUPAC Name ((2R,5S)-5-(4-amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolan-2-yl)methyl trihydrogen diphosphate triethylamine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 508.4
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.