Learn More
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Product Code. 12794868 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
EMT Duo Torch
For Use With (Equipment):
iCAP 7000 Series
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
12794868 EMT Duo Torch iCAP 7000 Series Each
1 options
Product Code. 12794868 Supplier Thermo Scientific™ Supplier No. 842312051841
This item requires a price update prior to being ordered.
Request a Price for this item here.
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance plasma torch for iCAP™ Series ICP-OES systems.
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for Thermo Scientific™ iCAP™ Series ICP-OES systems. The torch clips in and can be removed in just seconds.
Enhanced performance through advanced accessories We offer a dedicated torch designs for radial and axial plasma views, including:
  • EMT Radial Torch for radial-only systems
  • EMT Duo Torch for radial and axial (duo) view systems
Specifications
For Use With (Equipment) iCAP 7000 Series
Type EMT Duo Torch