missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMSY Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £366.00
Specifications
| Antigen | EMSY |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18346906
|
Novus Biologicals
NBP3-03289-20UL |
20 μL |
£153.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18343714
|
Novus Biologicals
NBP3-03289-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EMSY Polyclonal specifically detects EMSY in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| EMSY | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| chromosome 11 open reading frame 30, EMSYFLJ90741, protein EMSY | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1153-1322 of human EMSY (NP_064578.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| PBS with 50% glycerol, pH7.3. | |
| 56946 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title