missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Emricasan

Product Code. 18014902
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18014902

Brand: Tocris Bioscience™ 7310/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent pan-caspase inhibitor

Emricasan is a potent caspase inhibitor (IC50 values are 0.3, 0.4, 2, 4, 6, 6 and 20 nM for caspase-9, -1, -3, -6, -7, -8, and -2, respectively). Emricasan is antiapoptotic by inhibiting caspase 8. It also preserves cell viability after infection with several strains of Zika. Emricasan promotes survival of pluripotent stem cells in culture when used in combination with Chroman-1 (Cat. No. 7163), trans-ISRIB (Cat. No. 5284) and Polyamine Supplement x1000 (lyophilized) (Cat. No. 7739), a combination known as CEPT. When used as part of the CEPT cocktail, Emricasan improves survival of differentiated cells following cryopreservation. The compound also reduces hepatic inflammation in an animal model of liver injury. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-(1,1-Dimethylethyl)phenyl]-2-oxoglycyl-N-[(1S)-1-(carboxymethyl)-2-oxo-3-(2,3,5,6-tetrafluorophenoxy)propyl]-L-alaninamide
CAS 254750-02-2
Quantity 25 mg
Target Caspase Inhibitors
Molecular Formula C26H27F4N3O7
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.