missing translation for 'onlineSavingsMsg'
Learn More

Emricasan, MedChemExpress

Product Code. 16444946
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444946 100 mg 100mg
16440277 50 mg 50mg
16436637 5 mg 5mg
16433918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444946 Supplier Medchem Express Supplier No. HY10396
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Emricasan (PF 03491390) is an orally active and irreversible pan-caspase inhibitor. Emricasan inhibits Zika virus (ZIKV)-induced increases in caspase-3 activity and protected human cortical neural progenitors.

Emricasan (PF 03491390) is an orally active and irreversible pan-caspase inhibitor. Emricasan inhibits Zika virus (ZIKV)-induced increases in caspase-3 activity and protected human cortical neural progenitors.

Specifications

Chemical Name or Material Emricasan
CAS 254750-02-2
Color White
Purity Grade Notes Research
Molecular Formula C26H27F4N3O7
Quantity 100 mg
Synonym PF 03491390 IDN-6556
Solubility Information DMSO : 100 mg/mL (175.59 mM; Need ultrasonic)
SMILES FC1=C(C(F)=C(C=C1F)F)OCC([C@@H](NC([C@@H](NC(C(NC2=C(C=CC=C2)C(C)(C)C)=O)=O)C)=O)CC(O)=O)=O
Molecular Weight (g/mol) 569.5
Formula Weight 569.5
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.