missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Emricasan

Product Code. 18055742
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18055742 5 mg 5mg
18014902 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18055742 Supplier Tocris Bioscience™ Supplier No. 7310/5
  • ONLINE EXCLUSIVE PRICE
    £135.00
    £127.05 / 5mg
    Save £7.95
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Potent pan-caspase inhibitor

Emricasan is a potent caspase inhibitor (IC50 values are 0.3, 0.4, 2, 4, 6, 6 and 20 nM for caspase-9, -1, -3, -6, -7, -8, and -2, respectively). Emricasan is antiapoptotic by inhibiting caspase 8. It also preserves cell viability after infection with several strains of Zika. Emricasan promotes survival of pluripotent stem cells in culture when used in combination with Chroman-1 (Cat. No. 7163), trans-ISRIB (Cat. No. 5284) and Polyamine Supplement x1000 (lyophilized) (Cat. No. 7739), a combination known as CEPT. When used as part of the CEPT cocktail, Emricasan improves survival of differentiated cells following cryopreservation. The compound also reduces hepatic inflammation in an animal model of liver injury. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-(1,1-Dimethylethyl)phenyl]-2-oxoglycyl-N-[(1S)-1-(carboxymethyl)-2-oxo-3-(2,3,5,6-tetrafluorophenoxy)propyl]-L-alaninamide
CAS 254750-02-2
Quantity 5 mg
Target Caspase Inhibitors
Molecular Formula C26H27F4N3O7
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.