missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EMR3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13444419
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13444419 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13444419 Supplier Invitrogen™ Supplier No. PA532747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%); Orangutan, Gibbon (94%); Elephant (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

EMR3 gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 2 on chromosome 19. This protein may play a role in myeloid-myeloid interactions during immune and inflammatory responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMR3
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRE3
Gene Accession No. Q9BY15
Gene Alias ADGRE3; Adhesion G protein-coupled receptor E3; egf-like module containing, mucin-like, hormone receptor-like 3; EGF-like module receptor 3; EGF-like module-containing mucin-like hormone receptor-like 3; egf-like module-containing mucin-like receptor 3; EMR3; UNQ683/PRO1562
Gene Symbols ADGRE3
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminal cytoplasmic domain of human EMR3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84658
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.