missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emr1, Rat anti-Mouse, APC, Clone: BM8, Abnova™
Rat Monoclonal Antibody
Brand: Abnova MAB15511.100ug
This item is not returnable.
View return policy
Specifications
| Emr1 | |
| Monoclonal | |
| APC | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| DD7A5-7/EGF-TM7/F4/80/Gpf480/Ly71/TM7LN3 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| BM8 | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| EGF-like module containing, mucin-like, hormone receptor-like sequence 1 | |
| Emr1 | |
| Murine macrophages. | |
| RUO | |
| 13733 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction