missing translation for 'onlineSavingsMsg'
Learn More

EMR1 Antibody, Novus Biologicals™

Product Code. 18354758 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354758 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354758 Supplier Novus Biologicals Supplier No. H00002015B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

EMR1 Polyclonal antibody specifically detects EMR1 in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMR1
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH59395
Gene Alias ADGRE1, egf-like module containing, mucin-like, hormone receptor-like 1, egf-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EMR1 hormone receptor, TM7LN3
Host Species Mouse
Immunogen EMR1 (AAH59395, 1 a.a. - 821 a.a.) full-length human protein. MRGFNLLLFWGCCVMHSWEGHIRPTRKPNTKGNNCRDSTLCPAYATCTNTVDSYYCACKQGFLSSNGQNHFKDPGVRCKDIDECSQSPQPCGPNSSCKNLSGRYKCSCLDGFSSPTGNDWVPGKPGNFSCTDINECLTSSVCPEHSDCVNSMGSYSCSCQVGFISRNSTCEDVDECADPRACPEHATCNNTVGNYSCFCNPGFESSSGHLSFQGLKASCEDIDECTEMCPINSTCTNTPGSYFCTCHPGFAPSNGQLNFTDQGVECRDIDECRQDPSTCGPNSICTNALGSYSCGCIAGFHPNPEGSQKDGNFSCQRVLFKCKEDVIPDNKQIQQCQEGTAVKPAYVSFCAQINNIFSVLDKVCENKTTVVSLKNTTESFVPVLKQISTWTKFTKEETSSLATVFLESVESMTLASFWKPSANITPAVRTEYLDIESKVINKECSEENVTLDLVAKGDKMKIGCSTIEESESTETTGVAFVSFVGMESVLNERFFKDHQAPLTTSEIKLKMNSRVVGGIMTGEKKDGFSDPIIYTLENIQPKQKFERPICVSWSTDVKGGRWTSFGCVILEASETYTICSCNQMANLAIIMASGELTMGCAIIAGFLHYLFLACFFWMLVEAVILFLMVRNLKVVNYFSSRNIKMLHICAFGYGLPMLVVVISASVQPQGYGMHNRCWLNTETGFIWSFLGPVCTVIVINSLLLTWTLWILRQRLSSVNAEVSTLKDTRLLTFKAFAQLFILGCSWVLGIFQIGPVAGVMAYLFTIINSLQGAFIFLIHCLLNGQVREEYKRWITGKTKPSSQSQTSRILLSSMPSASKTG
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2015
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.