missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Empty Optimus Rack for CleanPak and Tip Eject Reloads
GREENER_CHOICE

Product Code. 30216550
Change view
Click to view available options
Size:
10 to 300 μL
1000 to 1250 μL
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30216550 1000 to 1250 μL Pack of 10
30216529 10 to 300 μL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30216550 Supplier BIOTIX™ Supplier No. 63300152
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

BIOTIX™ Empty Optimus Rack for CleanPak and Tip Eject Reloads is compatible with CleanPak and TipEject reloads for easy transfer and racking

  • Locking Clasp to secure tips during transport
  • Full 180 degree hinge design for convenient multichannel pipetting
  • SBS footprint can be used as a 96 well automation reservoir with pyramidal wells
  • Fully autoclaveable for multiple uses with CleanPak and Tip Eject reloads

Order Info

10 racks/pack

Certifications

ISO 9001:2015, Certified free of DNASE, RNASE, Endotoxins/pyrogens, PCR inhibitors. Sterilized product is certified to 10^-3 SAL.

TRUSTED_SUSTAINABILITY

Specifications

Description 1000 to 1250μL Optimus Empty Rack
Autoclavable Autoclavable
Color Clear
Holds Pipette Tips (20 to 300 μL, D300)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Application) uTIP and xTIP4 Reloads
For Use With (Equipment) CleanPak and TipEject
Quantity 10 Racks/Pk.
Size 1000 to 1250 μL
Length (Metric) 130 mm
Includes Base and lid
Product Type Empty Refill Rack with Lid
Array 8 x 12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.