missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Empasiprubart Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Empasiprubart Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283959
GREENER_CHOICE
View Documents
Invitrogen™
MA558841
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283314
View Documents
Invitrogen™
MA558842
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Empasiprubart, also known as ARGX-117, is a humanized monoclonal antibody specifically designed to bind and inhibit the function of the complement component 2 (C2) protein, a critical part of the complement cascade involved in immune responses. Complement component 2 (C2) is an essential part of the classical and lectin pathways of the complement system, pivotal in innate immunity. The C2 gene is located on chromosome 6p21.3 within the major histocompatibility complex (MHC) class III region. Structurally, C2 is a 100 kDa single-chain polypeptide that, in the presence of magnesium ions (Mg 2+), binds to C4b on the surface of target cells, forming part of the C3 convertase (C4b2a) complex. Upon activation by C1s, C2 is cleaved into C2a, the active enzyme of the convertase, and C2b. Functionally, C2a plays a critical role in catalyzing the conversion of C3 to C3a and C3b, leading to opsonization, inflammation, and eventual lysis of pathogens. Deficiency in C2 can lead to increased susceptibility to infections and autoimmune diseases. Structural studies suggest that C2's conformation changes upon binding with C4b, which is vital for its role in immune response.
Specifications

Specifications

Empasiprubart Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.