missing translation for 'onlineSavingsMsg'
Learn More

Empagliflozin, MedChemExpress

Product Code. 16486756
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486756 100 mg 100mg
16479077 5 mg 5mg
16431776 500 mg 500mg
16431519 1 g 1g
16426498 10 mg 10mg
16412597 50 mg 50mg
16401186 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16486756 Supplier Medchem Express Supplier No. HY15409
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Empagliflozin (BI 107730 is a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor with an IC50 of 3.1 nM for human SGLT-2.

Empagliflozin (BI 107730 is a selective sodium glucose cotransporter-2 (SGLT-2) inhibitor with an IC50 of 3.1 nM for human SGLT-2.

Specifications

Chemical Name or Material Empagliflozin
CAS 864070-44-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27ClO7
Quantity 100 mg
Synonym BI 10773
Solubility Information DMSO : 50 mg/mL (110.89 mM; Need ultrasonic) ∣H2O : 0.11 mg/mL (0.24 mM; Need ultrasonic and warming)
SMILES ClC(C=CC([C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)=C2)=C2CC(C=C3)=CC=C3O[C@H]4CCOC4
Molecular Weight (g/mol) 450.91
Formula Weight 450.91
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.