missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMP3 Polyclonal Antibody
Description
EMP3 Polyclonal Antibody for Western Blot
Probably involved in cell proliferation and cell-cell interactions.
Specifications
Specifications
| Antigen | EMP3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EMP3 |
| Gene Accession No. | P54852 |
| Gene Alias | Emp3; EMP-3; Epithelial membrane protein 3; H4; H-4; hematopoietic neural membrane protein 1; HNMP-1; MI-35; Protein YMP; YMP |
| Gene Symbols | EMP3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?