missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMP3 Monoclonal Antibody (439949)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561448
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Probably involved in cell proliferation and cell-cell interactions.
Specifications
| EMP3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P54852 | |
| EMP3 | |
| NS0 mouse myeloma cell line transfected with human EMP3, Met1-Glu163. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 439949 | |
| Unconjugated | |
| EMP3 | |
| Emp3; EMP-3; Epithelial membrane protein 3; H4; H-4; hematopoietic neural membrane protein 1; HNMP-1; MI-35; Protein YMP; YMP | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu