missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553220
This item is not returnable.
View return policy
Description
Immunogen sequence: DNAWWVGDEF FADVWRICTN NTNCTVINDS FQEYST Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 68%.
Epithelial membrane protein-2 (EMP2) is a member of the four transmembrane superfamily (TM4SF) and is thought to mediate trafficking of diverse proteins such as alpha6beta1 integrin and MHC class I to lipid raft microdomains. EMP2 has also recently been recognized as a putative tumor suppressor gene in certain model systems.
Specifications
| EMP2 | |
| Polyclonal | |
| Unconjugated | |
| EMP2 | |
| EMP2; EMP-2; Epithelial membrane protein 2; MGC9056; Protein XMP; Unknown (protein for MGC:137044); XMP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54851 | |
| EMP2 | |
| Recombinant protein corresponding to Human EMP2. Recombinant protein control fragment (Product #RP-91201). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction