missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMP/MAEA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17200
This item is not returnable.
View return policy
Description
EMP/MAEA Polyclonal specifically detects EMP/MAEA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| EMP/MAEA | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EMLP, EMPCell proliferation-inducing gene 5 protein, Erythroblast macrophage protein, HLC-10, Human lung cancer oncogene 10 protein, lung cancer-related protein 10, macrophage erythroblast attacher, PIG5, proliferation-inducing gene 5 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| Apoptosis | |
| 10296 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q7L5Y9 | |
| MAEA | |
| Recombinant protein encompassing a sequence within the center region of human MAEA. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction