missing translation for 'onlineSavingsMsg'
Learn More

EMP/MAEA Antibody [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30736907 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736907 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736907 Supplier Novus Biologicals Supplier No. AF7288I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

EMP/MAEA Polyclonal antibody specifically detects EMP/MAEA in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMP/MAEA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias EMLP, EMPCell proliferation-inducing gene 5 protein, Erythroblast macrophage protein, HLC-10, Human lung cancer oncogene 10 protein, lung cancer-related protein 10, macrophage erythroblast attacher, PIG5, proliferation-inducing gene 5
Host Species Sheep
Immunogen E. coli-derived recombinant human EMP/MAEA, Met12-Ser66, Accession # Q7L5Y9
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10296
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.