missing translation for 'onlineSavingsMsg'
Learn More

Emodin, Thermo Scientific™

A protein tyrosine kinase inhibitor

£90.50 - £485.00

Chemical Identifiers

CAS 518-82-1
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00001207
InChI Key RHMXXJGYXNZAPX-UHFFFAOYSA-N
Synonym emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
PubChem CID 3220
ChEBI CHEBI:42223
IUPAC Name 1,3,8-trihydroxy-6-methylanthracene-9,10-dione
SMILES CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404179
View Documents
Thermo Scientific Alfa Aesar
J61600.MC
100mg
£90.50
100mg
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15414179
View Documents
Thermo Scientific Alfa Aesar
J61600.MD
250mg
£177.00
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 11-09-2026
Add to basket
 
 Restricted Item
15494169
View Documents
Thermo Scientific Alfa Aesar
J61600.03
1g
£485.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 04-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

518-82-1
270.24
RHMXXJGYXNZAPX-UHFFFAOYSA-N
3220
1,3,8-trihydroxy-6-methylanthracene-9,10-dione
C15H10O5
MFCD00001207
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
CHEBI:42223
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O

Specifications

518-82-1
MFCD00001207
14,3561
RHMXXJGYXNZAPX-UHFFFAOYSA-N
1,3,8-trihydroxy-6-methylanthracene-9,10-dione
3220
270.24
C15H10O5
1888141
emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O
270.24
CHEBI:42223
Emodin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 208-258-8

missing translation for 'rtecsNumber' : CB7920600

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.