missing translation for 'onlineSavingsMsg'
Learn More

Emodin 8-Beta-D-Glucoside, TRC

Product Code. 30388275
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388275 1 mg
30373659 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388275 Supplier Toronto Research Chemicals Supplier No. TRCE5230101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23313-21-5
Molecular Formula C21 H20 O10
Molecular Weight (g/mol) 432.38
Synonym 1-(β-D-Glucopyranosyloxy)-3,8-dihydroxy-6-methyl-9,10-anthracenedione, 3,8-Dihydroxy-6-methyl-1-anthraquinonyl β-D-Glucopyranoside, 8-O-β-D-Glucopyranosyl Emodin, Anthraglycoside B, Emodin 8-O-β-D-Glucopyranoside, Emodin 8-O-β-D-Glucoside, Emodin 8-β-D-Glucopyranoside, Emodin Glucoside B, NSC 257449
IUPAC Name 1,6-dihydroxy-3-methyl-8-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyanthracene-9,10-dione
SMILES Cc1cc(O)c2C(=O)c3c(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)cc3C(=O)c2c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H20O10/c1-7-2-9-14(11(24)3-7)18(27)15-10(16(9)25)4-8(23)5-12(15)30-21-20(29)19(28)17(26)13(6-22)31-21/h2-5,13,17,19-24,26,28-29H,6H2,1H3/t13-,17-,19+,20-,21-/m1/s1
Formula Weight 432.1056
Chemical Name or Material Emodin 8-beta-D-Glucoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.