missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMMPRIN/CD147 Antibody (Centocor patent anti-CD147) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28146JF525
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| EMMPRIN/CD147 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| EMMPRIN/ CD147 | |
| 0.1 mL | |
| Cancer, Signal Transduction, Vision | |
| 682 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Centocor patent anti-CD147 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction