missing translation for 'onlineSavingsMsg'
Learn More

EMMPRIN/CD147 Antibody (125), DyLight 680, Novus Biologicals™

Product Code. 18322250 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322250 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322250 Supplier Novus Biologicals Supplier No. NBP289320FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMMPRIN/CD147
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 125
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EMMPRIN/CD147 (NP_940991.1; Met 1-His 205).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 682
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.