missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMMPRIN/CD147 Antibody (125), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89320B
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| EMMPRIN/CD147 | |
| Monoclonal | |
| Biotin | |
| 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EMMPRIN/CD147 (NP_940991.1; Met 1-His 205). | |
| 0.1 mL | |
| Cancer, Signal Transduction, Vision | |
| 682 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 125 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction