missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ EMM, Powdered Media
Shop All MP Biomedicals Products
Click to view available options
Quantity:
227 g
454 g
1000 g
Unit Size:
1kg
227g
454g
Description
Formulation
- Contents/liter: 3 g phthalic Acid K+, 2.2 g Na2HPO4, 5 g NH4Cl, 20 g dextrose
- Salts: 1.05 g MgCl2·6H2O, 14.7 μg CaCl2·2H2O, 1 g KCl, 0.04 g Na2SO4
- Vitamins: 1 mg pantothenic acid, 10 mg nicotinic acid, 10 mg myo-inositol, 10 μg biotin
- Minerals: 0.5 mg boric acid, 0.4 mg MnSO4, 0.4 mg ZnSO4·7H2O, 0.2 mg FeCl2·6H2O, 40 μg molybdic acid, 0.1 mg KI, 40 μg CuSO4·5H20, 1 mg citric acid
Applications:
Yeast cell culture, Life sciences
Specifications
Specifications
| Certifications/Compliance | Certified Thiamine (Vitamin B1) free |
| Content And Storage | 15° to 30°C |
| Form | Powder |
| Grade | Molecular biology |
| Product Type | EMM |
| Formulation | phthalic Acid K+ 3 g/L, Na2HPO4 2.2 g/L, NH4Cl 5 g/L, Dextrose 20 g/L, MgCl2.6H2O 1.05 g/L, CaCl2.2H2O 14.7 μg/L, KCl 1 g/L, Na2SO4 0.04 g/L, Pantothenic Acid 1 mg/L, Nicotinic Acid 10 mg/L, myo-inositol 10 mg/L, Biotin 10 μg/L, Boric Acid 0.5 mg/L, MnSO4 0.4 mg/L, ZnSO4.7H2O 0.4 mg/L, FeCl2.6H2O 0.2 mg/L, Molybdic Acid 40 μg/L, KI 0.1 mg/L, CuSO4.5H2O 40 μg/L, Citric Acid 1 mg/L |
| For Use With (Application) | For growth of fission Yeast S. Pombe |
| Quantity | 1000 g |
Safety and Handling
Product Identifier
- EMM Broth (Powder)
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Ammonium Chloride, Sodium Phosphate Dibasic Anhydrous
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction